Invitrogen
Manufacturer Code:PA526126
Catalog # PIPA526126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from human ATG1 conjugated to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATG1; ATG1 ATG1A UNC51 Unc51.1 hATG1 ATG1 autophagy related 1 homolog autophagy-related protein 1 homolog serine/threonine-protein kinase ULK1; ATG1 autophagy related 1 homolog; Autophagy-related protein 1 homolog; Serine/threonine-protein kinase ULK1; Serine/threonine-protein kinase Unc51.1; Unc-51-like kinase 1
Gene Aliases: ATG1; ATG1A; AU041434; hATG1; KIAA0722; mKIAA0722; ULK1; UNC51; Unc51.1
UniProt ID: (Human) O75385, (Mouse) O70405
Entrez Gene ID: (Human) 8408, (Mouse) 22241
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.