Invitrogen
Manufacturer Code:PA577810
Catalog # PIPA577810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from the mid-protein of Human ULK1 (aa 567-577) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATG1; ATG1 autophagy related 1 homolog; Autophagy-related protein 1 homolog; Serine/threonine-protein kinase ULK1; Serine/threonine-protein kinase Unc51.1; Unc-51 like kinase 1 (C. elegans) Autophagy related protein 1 homolog Autophagy-related protein 1 homolog FLJ38455 Serine/threonine-protein kinase ULK1 Unc51.1 ATG1A UNC51 C. elegans homolog of EC:2.7.11.1 Serine/threonine protein kinase Unc51.1 FLJ46475 ATG 1 hATG1 ATG1 Unc-51-like kinase 1 Serine/threonine protein kinase ULK1 ULK1 Unc 51 like kinase 1 KIAA0722 UNC 51 ULK 1 ULK1&lowbarHUMAN UNC51 Unc 51 (C. elegans) like kinase 1 ATG1 autophagy related 1 homolog; Unc-51-like kinase 1
Gene Aliases: ATG1; ATG1A; AU041434; hATG1; KIAA0722; mKIAA0722; ULK1; UNC51; Unc51.1
UniProt ID: (Human) O75385, (Mouse) O70405
Entrez Gene ID: (Human) 8408, (Rat) 360827, (Mouse) 22241
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.