Proteintech
Manufacturer Code:177151AP150UL
Catalog # 50-173-1737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ULBP1 Fusion Protein Ag12079 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALCAN beta N2DL 1 N2DL1 NKG2D ligand 1 NKG2DL1 RAET1I UL16 binding protein 1 ULBP1; ALCAN-beta; N2DL-1; NKG2D ligand 1; Retinoic acid early transcript 1I; UL16-binding protein 1; UL16-binding protein-like transcript 1
Gene Aliases: A430108B07Rik; MULT1; N2DL-1; N2DL1; NKG2DL1; RAET1I; ULBP1
UniProt ID: (Human) Q9BZM6
Entrez Gene ID: (Human) 80329, (Mouse) 77777
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.