Invitrogen
Manufacturer Code:PA527969
Catalog # PIPA527969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 51 and 516 of Human UHRF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase UHRF1; hUHRF1; HuNp95; ICBP90 Np95 RNF106 hNP95 E3 ubiquitin-protein ligase UHRF1 RING finger protein 106 huNp95 inverted CCAAT box-binding protein of 90 kDa nuclear protein 95 nuclear zinc finger protein Np95 transcription factor ICBP90 ubiquitin-like PHD and RING finger domain-containing protein 1 ub; Inverted CCAAT box-binding protein of 90 kDa; nuclear phosphoprotein 95; Nuclear protein 95; Nuclear zinc finger protein Np95; RING finger protein 106; RING-type E3 ubiquitin transferase UHRF1; Transcription factor ICBP90; Ubiquitin-like PHD and RING finger domain-containing protein 1; ubiquitin-like with PHD and ring finger domains 1; Ubiquitin-like-containing PHD and RING finger domains protein 1
Gene Aliases: hNP95; hUHRF1; huNp95; ICBP90; NP95; RNF106; TDRD22; UHRF1
UniProt ID: (Human) Q96T88
Entrez Gene ID: (Human) 29128
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.