Abnova
Manufacturer Code:H00029128A01
Catalog # 89-106-785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 694 a.a. &bsim 793 a.a) partial recombinant protein with GST tag.,UHRF1 (NP_037414 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant UHRF1.
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase UHRF1; FLJ21925; hNP95; hUHRF1; HuNp95; ICBP90; Inverted CCAAT box-binding protein of 90 kDa; MGC138707; Np95; nuclear phosphoprotein 95; Nuclear protein 95; Nuclear zinc finger protein Np95; RING finger protein 106; RING-type E3 ubiquitin transferase UHRF1; RNF106; Transcription factor ICBP90; Ubiquitin-like PHD and RING finger domain-containing protein 1; ubiquitin-like with PHD and ring finger domains 1; Ubiquitin-like-containing PHD and RING finger domains protein 1
Gene Aliases: hNP95; hUHRF1; huNp95; ICBP90; NP95; RNF106; TDRD22; UHRF1
UniProt ID: (Human) Q96T88
Entrez Gene ID: (Human) 29128
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.