Invitrogen
Manufacturer Code:PA537282
Catalog # PIPA537282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human UGT8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-hydroxyacylsphingosine 1-beta-galactosyltransferase; 2-hydroxyacylsphingosine 1-beta-galactosyltransferase Ceramide UDP-galactosyltransferase Cerebroside synthase UDP-galactose-ceramide galactosyltransferase CGT UGT4; Ceramide UDP-galactosyltransferase; Cerebroside synthase; CGalT; UDP galactosyltransferase 8; UDP galactosyltransferase 8A; UDP-galactose ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase 8; UDP-galactose-ceramide galactosyltransferase 8A; UDP-glucuronosyltransferase 8; uridine diphosphate glycosyltransferase 8
Gene Aliases: AI850488; AW455908; CGT; mCerGT; UGT4; UGT8; Ugt8a
UniProt ID: (Human) Q16880, (Rat) Q09426, (Mouse) Q64676
Entrez Gene ID: (Human) 7368, (Rat) 50555, (Mouse) 22239
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.