Invitrogen
Manufacturer Code:PA5103177
Catalog # PIPA5103177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human UGT8 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-hydroxyacylsphingosine 1-beta-galactosyltransferase; 2-hydroxyacylsphingosine 1-beta-galactosyltransferase AI850488 AW455908 ceramide UDP-galactosyltransferase Cerebroside synthase Cgt mCerGT UDP galactosyltransferase 8 UDP galactosyltransferase 8A UDP glycosyltransferase 8 UDP-galactose ceramide galactosyltransferase UDP-galactose-ceramide galactosyltransferase UDP-galactose-ceramide galactosyltransferase 8 UDP-galactose-ceramide galactosyltransferase 8A UDP-glucuronosyltransferase 8 UGT4 Ugt8 Ugt8a uridine diphosphate glycosyltransferase 8; Ceramide UDP-galactosyltransferase; Cerebroside synthase; CGalT; UDP galactosyltransferase 8; UDP galactosyltransferase 8A; UDP-galactose ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase 8; UDP-glucuronosyltransferase 8; uridine diphosphate glycosyltransferase 8
Gene Aliases: CGT; UGT4; UGT8; Ugt8a
UniProt ID: (Human) Q16880, (Rat) Q09426
Entrez Gene ID: (Human) 7368, (Rat) 50555
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.