Invitrogen
Manufacturer Code:PA523863
Catalog # PIPA523863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 377-405 amino acids from the C-terminal region of human UGT3A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: UDP glucuronosyltransferase 3 family, polypeptide A2; UDP glycosyltransferase 3 family, polypeptide A2; UDP-glucuronosyltransferase 3A2; UDP-glucuronosyltransferase 3A2 UDPGT 3A2; UDPGT 3A2
Gene Aliases: AI313915; PSEC0073; UGT3A2; UNQ842/PRO1780
UniProt ID: (Human) Q3SY77, (Mouse) Q8JZZ0
Entrez Gene ID: (Human) 167127, (Mouse) 223337
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.