Novus Biologicals
Manufacturer Code:NBP293386100UL
Catalog # NB028654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 23-300 of human UGT3A2 (NP_777574.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.1.17 MGC119426 MGC119429 UDP glycosyltransferase 3 family polypeptide A2 UDP-glucuronosyltransferase 3A2 UDPGT 3A2; UDP glycosyltransferase 3 family, polypeptide A2; UDP-glucuronosyltransferase 3A2; UDPGT 3A2
Gene Aliases: PSEC0073; UGT3A2; UNQ842/PRO1780
UniProt ID: (Human) Q3SY77
Entrez Gene ID: (Human) 167127
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.