Novus Biologicals
Manufacturer Code:NBP288555
Catalog # NB036177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human UGT3A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.1 EC 2.4.1.17 FLJ26528 FLJ34658 UDP glycosyltransferase 3 family polypeptide A1 UDP-glucuronosyltransferase 3A1 UDPGT 3A1; UDP glycosyltransferase 3 family, polypeptide A1; UDP-glucuronosyltransferase 3A1; UDPGT 3A1
Gene Aliases: UGT3A1
UniProt ID: (Human) Q6NUS8
Entrez Gene ID: (Human) 133688
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.