Proteintech
Manufacturer Code:166611AP150UL
Catalog # 50-173-1094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UGT2B7 Fusion Protein Ag10121 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3,4-catechol estrogen specific; 3,4-catechol estrogen-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B7; UDP glucuronosyltransferase 2B7; UDP-glucuronosyltransferase 2B7; UDP-glucuronosyltransferase 2B9; UDP-glucuronyltransferase, family 2, beta-7; UDPGT 2B7; UDPGT 2B7 UDPGT 2B9 UDPGTh 2 UGT2B7 UGT2B9 UGTB2B9; UDPGT 2B9; UDPGTh-2
Gene Aliases: UDPGT 2B9; UDPGT2B7; UDPGTH2; UGT2B7; UGT2B9; UGTB2B9
UniProt ID: (Human) P16662
Entrez Gene ID: (Human) 7364
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.