Novus Biologicals
Manufacturer Code:NBP169604
Catalog # NBP169604
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to UGT2B4(UDP glucuronosyltransferase 2 family polypeptide B4) The peptide sequence was selected from the N terminal of UGT2B4. Peptide sequence NIKTILDELVQRGHEVTVLASSASISFDPNSPSTLKFEVYPVSLTKTEFE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: family 2 beta-4 UDP glucuronosyltransferase 2 family polypeptide B4 UDP glycosyltransferase 2 family polypeptide B4; HLUG25; hyodeoxycholic acid; Hyodeoxycholic acid-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B4; UDP-glucuronosyltransferase 2B4; UDP-glucuronyltransferase, family 2, beta-4; UDPGT 2B4; UDPGTh-1
Gene Aliases: HLUG25; UDPGT2B4; UDPGTh-1; UDPGTH1; UGT2B11; UGT2B4
UniProt ID: (Human) P06133
Entrez Gene ID: (Human) 7363
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.