Abnova
Manufacturer Code:H00007363K
Catalog # 89-203-398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human UGT2B4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human UGT2B4 peptide using ARM Technology.
For Research Use Only
Protein Aliases: HLUG25; hyodeoxycholic acid; Hyodeoxycholic acid-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B4; UDP-glucuronosyltransferase 2B4; UDP-glucuronyltransferase, family 2, beta-4; UDPGT 2B4; UDPGTh-1; UGT2B11
Gene Aliases: HLUG25; UDPGT2B4; UDPGTh-1; UDPGTH1; UGT2B11; UGT2B4
UniProt ID: (Human) P06133
Entrez Gene ID: (Human) 7363
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.