Invitrogen
Manufacturer Code:PA513430
Catalog # PIPA513430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 119-147 amino acids from the central region of human UDB17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C19-steroid-specific UDP-glucuronosyltransferase; C19-steroid-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B17; UDP glycosyltransferase 2 family, member B17; UDP-glucuronosyltransferase 2B17; UDP-glucuronosyltransferase 2B17 UDPGT 2B17 C19-steroid-specific UDP-glucuronosyltransferase UGT2B17; UDP-glucuronyltransferase, family 2, beta-17; UDPGT 2B17
Gene Aliases: BMND12; UDPGT2B17; UGT2B17
UniProt ID: (Human) O75795
Entrez Gene ID: (Human) 7367
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.