Novus Biologicals
Manufacturer Code:NBP294747
Catalog # NB024014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 60-190 of human UGT2B15 (NP_001067.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.1 EC 2.4.1.17 HLUG4 UDP glucuronosyltransferase 2 family polypeptide B15 UDP glycosyltransferase 2 family polypeptide B15 UDP glycosyltransferase 2B15 UDP-glucuronosyltransferase 2B15 UDP-glucuronosyltransferase 2B8 UDP-glucuronosyltransferase UGT2B15 UDP-glucuronyltransferase family 2 beta-15 UDPGT 2B15 UDPGT 2B8 UDPGTh-3 UDPGTH3 UGT2B8UDPGT2B15; HLUG4; UDP glucuronosyltransferase 2 family, polypeptide B15; UDP glycosyltransferase 2B15; UDP-glucuronosyltransferase 2B15; UDP-glucuronosyltransferase 2B8; UDP-glucuronosyltransferase UGT2B15; UDP-glucuronyltransferase, family 2, beta-15; UDPGT 2B15; UDPGT 2B8; UDPGTh-3
Gene Aliases: HLUG4; UDPGT 2B8; UDPGT2B15; UDPGTH3; UGT2B15; UGT2B8
UniProt ID: (Human) P54855
Entrez Gene ID: (Human) 7366
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.