Invitrogen
Manufacturer Code:PA522329
Catalog # PIPA522329
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 291 and 530 of UGT1A9 (Uniprot IDNo.O60656) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HLUGP4 LUGP4 UDPGT UDPGT 1-9 UGT-1I UGT1-09 UGT1-9 UGT1.9 UGT1AI UGT1I UDP glycosyltransferase 1 family polypeptide A9 UDP-glucuronosyltransferase 1-9 UDP-glucuronosyltransferase 1-I UDP-glucuronosyltransferase 1A9; lugP4; UDP glucuronosyltransferase 1 family, polypeptide A9; UDP glycosyltransferase 1 family polypeptide A12; UDP glycosyltransferase 1 family, polypeptide A9; UDP-glucuronosyltransferase 1 family polypeptide A9s; UDP-glucuronosyltransferase 1-7; UDP-glucuronosyltransferase 1-9; UDP-glucuronosyltransferase 1-I; UDP-glucuronosyltransferase 1A9; UDPGT; UDPGT 1-9; UGT-1I; UGT1A12; UGT1A9; UGTP4
Gene Aliases: GNT1; HLUGP4; LUGP4; mUGTBr/P; UDPGT; UDPGT 1-9; UGT-1I; UGT1; UGT1-09; UGT1-9; UGT1.9; Ugt1a12; UGT1A9; UGT1A9S; UGT1AI; UGT1I; UGTP4
UniProt ID: (Human) O60656, (Mouse) Q62452
Entrez Gene ID: (Human) 54600, (Mouse) 394434
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.