Abnova
Manufacturer Code:H00054576K
Catalog # 89-201-707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human UGT1A8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human UGT1A8 peptide using ARM Technology.
For Research Use Only
Protein Aliases: UDP glucuronosyltransferase 1 family, polypeptide A8; UDP glycosyltransferase 1 family, polypeptide A8; UDP-glucuronosyltransferase 1 family polypeptide A8s; UDP-glucuronosyltransferase 1-8; UDP-glucuronosyltransferase 1-H; UDP-glucuronosyltransferase 1A8; UDPGT 1-8; UDPGT UGT1A8S UGT1H; UGT-1H; UGT1A8
Gene Aliases: GNT1; UDPGT; UDPGT 1-8; UGT-1H; UGT1; UGT1-08; UGT1.8; UGT1A8; UGT1A8S; UGT1H
UniProt ID: (Human) Q9HAW9
Entrez Gene ID: (Human) 54576
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.