Invitrogen
Manufacturer Code:PA522319
Catalog # PIPA522319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 8 and 227 of UGT1A6 (Uniprot IDNo.P19224) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GNT1 HLUGP HLUGP1 UDPGT UDPGT 1-6 UGT1 UGT1A6S UGT1F UDP glycosyltransferase 1 family polypeptide A6 UDP-glucuronosyltransferase 1 family polypeptide A6s UDP-glucuronosyltransferase 1-6 UDP-glucuronosyltransferase 1-F UDP-glucuronosyltransferase 1A6 UGT-1F UGT1*6 UGT1-06 UGT1.6 phenol-metabolizing UDP-glucuronosyltransferase; Phenol UDP-glucuronosyltransferase; Phenol-metabolizing UDP-glucuronosyltransferase; UDP glucuronosyltransferase 1 family, polypeptide A6; UDP glycosyltransferase 1 family, polypeptide A6; UDP-glucuronosyltransferase 1 family polypeptide A6s; UDP-glucuronosyltransferase 1-6; UDP-glucuronosyltransferase 1-F; UDP-glucuronosyltransferase 1A6; UDPGT 1-6; UGP1A1; UGT-1F; UGT1*6; UGT1-06; UGT1.6; UGT1A6; UGT1A7
Gene Aliases: GNT1; HLUGP; HLUGP1; UDPGT; UDPGT 1-6; UGT1; UGT1-06; UGT1.6; UGT1A6; Ugt1a6a; UGT1A6S; Ugt1a7; UGT1F
UniProt ID: (Human) P19224, (Mouse) Q64435
Entrez Gene ID: (Human) 54578, (Mouse) 94284
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.