Novus Biologicals
Manufacturer Code:NBP133743
Catalog # NBP133743
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human UGT1A6. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GNT1UDPGT 1-6 HLUGP HLUGP1 MGC29860 Phenol-metabolizing UDP-glucuronosyltransferase UDP glucuronosyltransferase 1 family polypeptide A6 UDP glycosyltransferase 1 family polypeptide A6 UDP-glucuronosyltransferase 1 family polypeptide A6s UDP-glucuronosyltransferase 1-6 UDP-glucuronosyltransferase 1A6 UDP-glucuronosyltransferase 1-F UDPGT UGT1 UGT1*6 UGT1.6 UGT1-06 UGT1A6S UGT-1F UGT1FEC 2.4.1.17; Phenol-metabolizing UDP-glucuronosyltransferase; UDP glucuronosyltransferase 1 family, polypeptide A6; UDP glycosyltransferase 1 family, polypeptide A6; UDP-glucuronosyltransferase 1 family polypeptide A6s; UDP-glucuronosyltransferase 1-6; UDP-glucuronosyltransferase 1-F; UDP-glucuronosyltransferase 1A6; UDPGT 1-6; UGT-1F; UGT1*6; UGT1-06; UGT1.6
Gene Aliases: GNT1; HLUGP; HLUGP1; UDPGT; UDPGT 1-6; UGT1; UGT1A6; UGT1A6S; UGT1F
UniProt ID: (Human) P19224
Entrez Gene ID: (Human) 54578
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.