Invitrogen
Manufacturer Code:PA590760
Catalog # PIPA590760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UGT1A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bilirubin UDP-glucuronosyltransferase isozyme 2; Bilirubin-specific UDPGT isozyme 2; hUG-BR2; UDP glucuronosyltransferase 1 family, polypeptide A4; UDP glycosyltransferase 1 family, polypeptide A4; UDP-glucuronosyltransferase 1 family polypeptide A4s; UDP-glucuronosyltransferase 1-4; UDP-glucuronosyltransferase 1-D; UDP-glucuronosyltransferase 1A4; UDPGT 1-4; UGT-1D; UGT1A4
Gene Aliases: GNT1; HUG-BR2; UDPGT; UDPGT 1-4; UGT-1D; UGT1; UGT1-04; UGT1.4; UGT1A4; UGT1A4S; UGT1D
UniProt ID: (Human) P22310
Entrez Gene ID: (Human) 54657
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.