Invitrogen
Manufacturer Code:PA589315
Catalog # PIPA589315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-200 of human UGT1A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B1; bilirubin UDP-glucuronosyltranserase; bilirubin UDP-glucuronosyltransferase 1-1; bilirubin UDP-glucuronosyltransferase isozyme 1; Bilirubin-specific UDPGT isozyme 1; hUG-BR1; UDP glucuronosyltransferase 1 family, polypeptide A1; UDP glycosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1 family member 1; UDP-glucuronosyltransferase 1 family, member 1; UDP-glucuronosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1-A; UDP-glucuronosyltransferase 1A isoform 1; UDP-glucuronosyltransferase 1A1; UDPGT; UDPGT 1-1; UGT-1A; UGT1*1; UGT1-01; UGT1.1; UGT1A1; UGTBR1
Gene Aliases: BILIQTL1; GNT1; HUG-BR1; UDPGT; UDPGT 1-1; Udpgt-1a; UGT1; UGT1A; UGT1A01; UGT1A1; UgtBr1
UniProt ID: (Human) P22309, (Mouse) Q63886, (Rat) Q64550
Entrez Gene ID: (Human) 54658, (Mouse) 394436, (Rat) 24861
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.