Invitrogen
Manufacturer Code:PA576862
Catalog # PIPA576862
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human UGT1A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bilirubin specific UDPGT isozyme 1bilirubin UDP glucuronosyltransferase 1 1bilirubin UDP glucuronosyltransferase isozyme 1Bilirubin-specific UDPGT isozyme 1EC 2.4.1.17GNT1HUG BR1HUG-BR1HUGBR1PHENOL/BILIRUBIN UDP GLUCURONOSYLTRANSFERASEUD11&lowbarHUMANUDP glucuronosyltransferase 1 1 &lsqbPrecursor]UDP glucuronosyltransferase 1 family polypeptide A1UDP glucuronosyltransferase 1A1UDP GLYCOSYLTRANSFERASE 1UDP-glucuronosyltransferase 1-1UDP-glucuronosyltransferase 1-AUDP-glucuronosyltransferase 1A1UDPGTUDPGT 1-1UGT 1AUGT-1AUGT1UGT1 01UGT1*1UGT1-01UGT1.1UGT1AUgt1a1; bilirubin UDP-glucuronosyltranserase; bilirubin UDP-glucuronosyltransferase 1-1; bilirubin UDP-glucuronosyltransferase isozyme 1; Bilirubin-specific UDPGT isozyme 1; hUG-BR1; UDP glucuronosyltransferase 1 family, polypeptide A1; UDP glycosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1-A; UDP-glucuronosyltransferase 1A isoform 1; UDP-glucuronosyltransferase 1A1; UDPGT 1-1; UGT-1A; UGT1*1; UGT1-01; UGT1.1; UGT1A1
Gene Aliases: BILIQTL1; GNT1; HUG-BR1; UDPGT; UDPGT 1-1; UGT1; UGT1A; UGT1A1
UniProt ID: (Human) P22309
Entrez Gene ID: (Human) 54658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.