Novus Biologicals
Manufacturer Code:NBP180642
Catalog # NBP180642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RGAAKAIASASASASGPCVHQQPPGQPPPPQLSSHRGDLITEPFLPKLLTKVKGS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 35 (UDP-galactose transporter) member 2 solute carrier family 35 (UDP-galactose transporter) member A2 UDP-galactose translocator UGALTUGAT UGT1 UGT2UDP-galactose transporter UGTLSolute carrier family 35 member A2 UGTUDP-Gal-Tr; solute carrier family 35 (UDP-galactose transporter), member 2; solute carrier family 35 (UDP-galactose transporter), member A2; Solute carrier family 35 member A2; UDP-Gal-Tr; UDP-galactose translocator; UDP-galactose transporter
Gene Aliases: CDG2M; CDGX; SLC35A2; UDP-Gal-Tr; UGALT; UGAT; UGT; UGT1; UGT2; UGTL
UniProt ID: (Human) P78381
Entrez Gene ID: (Human) 7355
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.