Invitrogen
Manufacturer Code:PA527960
Catalog # PIPA527960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 690 and 1200 of Human UGCGL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HUGT2 UGCGL2 UGT2 UDP--Glc:glycoprotein glucosyltransferase 2 UDP-Glc:glycoprotein glucosyltransferase 2 UDP-glucose ceramide glucosyltransferase-like 1 UDP-glucose ceramide glucosyltransferase-like 2 UDP-glucose:glycoprotein glucosyltransferase 2; UDP--Glc:glycoprotein glucosyltransferase 2; UDP-Glc:glycoprotein glucosyltransferase 2; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose ceramide glucosyltransferase-like 2; UDP-glucose:glycoprotein glucosyltransferase 2; UGT2
Gene Aliases: HUGT2; UGCGL2; UGGT2; UGT2
UniProt ID: (Human) Q9NYU1
Entrez Gene ID: (Human) 55757
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.