Invitrogen
Manufacturer Code:PA5101950
Catalog # PIPA5101950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human UGDH corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDH UDP UGD UDPGDH UDP-Glc dehydrogenase UDP-GlcDH UDP-glucose 6-dehydrogenase UDP-glucose dehydrogeanse UDP-glucose dehydrogenase UGD UGDH uridine diphospho-glucose dehydrogenase; UDP-Glc dehydrogenase; UDP-GlcDH; UDP-glucose 6-dehydrogenase; UDP-glucose dehydrogeanse; UDP-glucose dehydrogenase; UDPGDH; uridine diphospho-glucose dehydrogenase
Gene Aliases: GDH; UDP-GlcDH; UDPGDH; UGD; UGDH
UniProt ID: (Human) O60701, (Rat) O70199, (Mouse) O70475
Entrez Gene ID: (Human) 7358, (Rat) 83472, (Mouse) 22235
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.