Invitrogen
Manufacturer Code:PA544978
Catalog # PIPA544978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human UFM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ubiquitin-fold modifier 1; Ubiquitin-fold modifier 1 BM-002 C13orf20 bA131P10.1; zBM-002
Gene Aliases: 1810045K17Rik; AI132708; AI463323; BM-002; C13orf20; ENSMUSG00000074598; Gm10726; RGD1304890; ubfm1; UFM1; wu:fa10f05; wu:fb72g01; zgc:55335
UniProt ID: (Bovine) Q2KJG2, (Human) P61960, (Mouse) Q3TLT0, (Rat) Q5BJP3, (Zebrafish) Q803Y4
Entrez Gene ID: (Bovine) 530547, (Dog) 477297, (Rabbit) 100357690, (Horse) 100066289, (Pig) 100522509, (Human) 51569, (Mouse) 67890, (Rat) 365797, (Zebrafish) 322756
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.