Novus Biologicals
Manufacturer Code:NBP198392
Catalog # NBP198392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is Uba5 - N-terminal region. Peptide sequence LLFDYDKVELANMNRLFFQPYQAGLSKVHAAEHTLRNINPDVLFEVHNYN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ17281 FLJ23251UBA5 ThiFP1 UBE1DC1ubiquitin-activating enzyme E1 homolog ubiquitin-like modifier activating enzyme 5 ubiquitin-like modifier-activating enzyme 5 UFM1-activating enzyme; ThiFP1; UBA5, ubiquitin-activating enzyme E1 homolog; Ubiquitin-activating enzyme 5; Ubiquitin-activating enzyme E1 domain-containing protein 1; ubiquitin-activating enzyme E1-domain containing 1; ubiquitin-like modifier activating enzyme 5; Ubiquitin-like modifier-activating enzyme 5; UFM1-activating enzyme
Gene Aliases: THIFP1; UBA5; UBE1DC1
UniProt ID: (Human) Q9GZZ9
Entrez Gene ID: (Human) 79876
Molecular Function: ligase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.