Abnova
Manufacturer Code:PAB22460
Catalog # 89-124-170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human UFL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant UFL1.
For Research Use Only
Protein Aliases: E3 UFM1-protein ligase 1; E3 UFM1-protein transferase 1; KIAA0776; LZAP-binding protein; Maxer; Multiple alpha-helix protein located at ER; NLBP; Novel LZAP-binding protein; RCAD; Regulator of C53/LZAP and DDRGK1; Regulator of CDK5RAP3 and DDRGK1; RP3-393D12.1; UFM1-specific ligase 1
Gene Aliases: KIAA0776; MAXER; NLBP; RCAD; UFL1
UniProt ID: (Human) O94874
Entrez Gene ID: (Human) 23376
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.