Novus Biologicals
Manufacturer Code:NBP103339
Catalog # NBP103339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 257 and 307 of human ubiquitin fusion degradation 1 using the numbering given in entry NP_005650.2 (GeneID 7353). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP1C ATP1G1ATPase Na+/K+ transporting gamma 1 polypeptide FXYD domain containing ion transport regulator 2 FXYD domain-containing ion transport regulator 2 HOMG2 hypomagnesemia 2 renal MGC12372 Na(+)/K(+) ATPase subunit gamma Sodium pump gamma chain sodium/potassium-transporting ATPase subunit gamma Sodium-potassium-ATPase gamma polypeptide; UB fusion protein 1; Ubiquitin fusion degradation protein 1; Ubiquitin recognition factor in ER-associated degradation protein 1
Gene Aliases: UFD1; UFD1L
UniProt ID: (Human) Q92890
Entrez Gene ID: (Human) 7353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.