Proteintech
Manufacturer Code:106151AP150UL
Catalog # 50-172-6450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UFD1L Fusion Protein Ag0954 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: UB fusion protein 1; UB fusion protein 1 UFD1 UFD1L; UB fusion protein 1 {ECO:0000250|UniProtKB:Q92890}; Ubiquitin fusion degradation protein 1; Ubiquitin fusion degradation protein 1 homolog; ubiquitin fusion degradation protein 1 homolog {ECO:0000250|UniProtKB:Q92890}; Ubiquitin recognition factor in ER-associated degradation protein 1; ufd1l {ECO:0000312|RGD:619822}
Gene Aliases: UFD1; UFD1L
UniProt ID: (Human) Q92890, (Rat) Q9ES53, (Mouse) P70362
Entrez Gene ID: (Human) 7353, (Rat) 84478, (Mouse) 22230
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.