Invitrogen
Manufacturer Code:PA555633
Catalog # PIPA555633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UFC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC155 Ufm1-conjugating enzyme 1 ubiquitin-fold modifier-conjugating enzyme 1; Ubiquitin-fold modifier-conjugating enzyme 1; Ufm1-conjugating enzyme 1; Ufm1-conjugating enzyme 1-
Gene Aliases: 1110021H02Rik; AA062310; CGI-126; ESTM29; HSPC155; UFC1; ufc1-s
UniProt ID: (Human) Q9Y3C8, (Rat) Q6BBI8, (Mouse) Q9CR09
Entrez Gene ID: (Human) 51506, (Rat) 445268, (Mouse) 66155
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.