Novus Biologicals
Manufacturer Code:NBP190906
Catalog # NBP190906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AVVICTEWDMFKELDYERIHKKMLKPAFIFDGRRVLDGLHNELQTIGFQIETIGKKVSSKRIPYAPSGEIPKFSLQDPPNKKPKV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.1.1.22 GDH UDPGDHUDP-glucose dehydrogenase UDP-Glc dehydrogenase UDP-GlcDH UDP-glucose 6-dehydrogenase UGD uridine diphospho-glucose dehydrogenase; UDP-Glc dehydrogenase; UDP-glucose 6-dehydrogenase; UDP-glucose dehydrogenase; uridine diphospho-glucose dehydrogenase
Gene Aliases: GDH; UDP-GlcDH; UDPGDH; UGD; UGDH
UniProt ID: (Human) O60701
Entrez Gene ID: (Human) 7358
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.