Abnova
Manufacturer Code:H00010331B01P
Catalog # 89-017-001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | B3GNT3 (AAH67423.1 1 a.a. ~ 372 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human B3GNT3 protein.
For Research Use Only
Protein Aliases: B3GAL-T8; B3GN-T3; B3GNT-3; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; Beta-1,3-galactosyltransferase 8; Beta-1,3-GalTase 8; beta-1,3-Gn-T3; beta-1,3-N-acetylglucosaminyltransferase bGnT-3; Beta-3-Gx-T8; beta3GalT8; beta3Gn-T3; BGnT-3; Core 1 extending beta-1,3-N-acetylglucosaminyltransferase; Core1-beta3GlcNAcT; HP10328; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 3; putative type II membrane protein; TMEM3; Transmembrane protein 3; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 8; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 8; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
Gene Aliases: B3GAL-T8; B3GALT8; B3GN-T3; B3GNT-3; B3GNT3; beta3Gn-T3; HP10328; TMEM3; UNQ637/PRO1266
UniProt ID: (Human) Q9Y2A9
Entrez Gene ID: (Human) 10331
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.