Abnova
Manufacturer Code:H00126792A01
Catalog # 89-004-778
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | B3GALT6 (NP_542172 229 a.a. ~ 329 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant B3GALT6.
For Research Use Only
Protein Aliases: Beta-1,3-galactosyltransferase 6; Beta-1,3-GalTase 6; beta3Gal-T6; beta3GalT6; GAG GalTII; Galactosyltransferase II; Galactosylxylosylprotein 3-beta-galactosyltransferase; UDP-Gal:betaGal beta 1,3-galactosyltransferase 6; UDP-Gal:betaGal beta 1,3-galactosyltransferase polypeptide 6; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 6
Gene Aliases: B3GALT6; beta3GalT6; EDSP2; SEMDJL1
UniProt ID: (Human) Q96L58
Entrez Gene ID: (Human) 126792
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.