Proteintech
Manufacturer Code:667001IG150UL
Catalog # 50-173-7116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B4B1 |
Immunogen | UCP2 Fusion Protein Ag19361 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BMIQ4 SLC25A8 UCP 2 UCP2 UCPH; Mitochondrial uncoupling protein 2; Solute carrier family 25 member 8; UCP 2; UCPH; uncoupling protein 2 (mitochondrial, proton carrier)
Gene Aliases: BMIQ4; SLC25A8; UCP2; UCPH
UniProt ID: (Human) P55851
Entrez Gene ID: (Human) 7351
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.