Invitrogen
Manufacturer Code:PA577553
Catalog # PIPA577553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GRRYQSTVEAYK corresponding to amino acid residues 153-164 of rat mitochondrial uncoupling protein 2 (UCP2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial uncoupling protein 2; Mitochondrial Uncoupling Protein 2 (UCP2); Solute carrier family 25 member 8; UCP 2; UCPH; uncoupling protein 2 (mitochondrial, proton carrier); Uncoupling protein 2, mitochondrial
Gene Aliases: BMIQ4; SLC25A8; UCP2; UCPH
UniProt ID: (Human) P55851, (Rat) P56500, (Mouse) P70406
Entrez Gene ID: (Human) 7351, (Rat) 54315, (Mouse) 22228
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.