R&D Systems
Manufacturer Code:AF4739SP
Catalog # AF4739SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human UCP2 Met1-Phe309 Accession # P55851 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial uncoupling protein 2; mitochondrial uncoupling protein 2 SLC25A8 SLC25A8BMIQ4 Solute carrier family 25 member 8 UCP 2 UCPH uncoupling protein 2 (mitochondrial proton carrier); Solute carrier family 25 member 8; UCP 2; UCPH; uncoupling protein 2 (mitochondrial, proton carrier)
Gene Aliases: BMIQ4; SLC25A8; UCP2; UCPH
UniProt ID: (Human) P55851
Entrez Gene ID: (Human) 7351
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.