Invitrogen
Manufacturer Code:PA552092
Catalog # PIPA552092
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UCKL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: UCK1-LIKE; uridine-cytidine kinase 1-like 1; Uridine-cytidine kinase-like 1; Uridine/cytidine kinase-like 1 UCKL1 URKL1
Gene Aliases: F538; UCK1L; UCKL1; URKL1
UniProt ID: (Human) Q9NWZ5
Entrez Gene ID: (Human) 54963
Molecular Function: glycosyltransferase kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.