Proteintech
Manufacturer Code:170051AP150UL
Catalog # 50-173-1324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UCKL1 Fusion Protein Ag10701 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: UCK1 LIKE UCK1L UCKL1 Uridine cytidine kinase like 1 URKL1; UCK1-LIKE; uridine kinase-like 1; uridine-cytidine kinase 1-like 1; Uridine-cytidine kinase-like 1
Gene Aliases: 1110007H10Rik; F538; RGD1565465; UCK1L; UCKL1; URKL1
UniProt ID: (Human) Q9NWZ5, (Mouse) Q91YL3
Entrez Gene ID: (Human) 54963, (Mouse) 68556, (Rat) 499956
Molecular Function: glycosyltransferase kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.