Novus Biologicals
Manufacturer Code:NBP274779PEATT594
Catalog # N274779P594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A12 |
Immunogen | Full length recombinant protein of human UCK (NP_113620) produced in HEK293T cells. |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: Cytidine monophosphokinase 1; Cytidine monophosphokinase 1 EC 2.7.1.48 FLJ12255 Uridine monophosphokinase 1 uridine-cytidine kinase 1 URK1UCK 1; UCK 1; Uridine monophosphokinase 1; Uridine-cytidine kinase 1
Gene Aliases: UCK1; URK1
UniProt ID: (Human) Q9HA47
Entrez Gene ID: (Human) 83549
Molecular Function: glycosyltransferase kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.