Abnova
Manufacturer Code:MAB10579
Catalog # 89-108-770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | YK-3E3 |
Immunogen | Recombinant protein corresponding to full length human UCHL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full-length recombinant S100A3.
For Research Use Only
Protein Aliases: Protein S-100E; Protein S100-A3; S100 calcium-binding protein A3; S100E
Gene Aliases: S100A3; S100E
UniProt ID: (Human) P33764
Entrez Gene ID: (Human) 6274
Molecular Function:
calcium-binding protein
calmodulin
growth factor
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.