Invitrogen
Manufacturer Code:PA552967
Catalog # PIPA552967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UBXN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2B28 SAKS1 UBXD10 SAPK substrate protein 1 UBA/UBX 33.3 kDa protein UBX domain-containing protein 1; mY33K; Protein 2B28; SAPK substrate protein 1; UBA/UBX 33.3 kDa protein; UBX domain-containing protein 1
Gene Aliases: 2B28; 4930455J02Rik; D19Ertd721e; RGD1309471; SAKS1; T25529; UBXD10; UBXN1
UniProt ID: (Human) Q04323, (Mouse) Q922Y1, (Rat) Q499N6
Entrez Gene ID: (Human) 51035, (Mouse) 225896, (Rat) 293719
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.