Novus Biologicals
Manufacturer Code:NBP198480
Catalog # NBP198480
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is Ubtd1 - C-terminal region. Peptide sequence LSASLPDTVGQLKRQLHSQEGIEPSWQRWFFSGKLLTDRTRLQETKIQKD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ11807 ubiquitin domain containing 1 ubiquitin domain-containing protein 1; Ubiquitin domain-containing protein 1
Gene Aliases: UBTD1
UniProt ID: (Human) Q9HAC8
Entrez Gene ID: (Human) 80019
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.