Novus Biologicals
Manufacturer Code:NB1001591
Catalog # NB1001591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 600 and 650 of human E3 Ubiquitin Protein Ligase HECTdomain containing 1 using the numbering given in entry NP_056986.2 (GeneID 51366) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DD5 E3 ubiquitin protein ligase HECT domain containing 1 E3 ubiquitin-protein ligase HECT domain-containing 1 EC 6.3.2 EC 6.3.2.- EDD1E3 ubiquitin-protein ligase UBR5 EDDE3 identified by differential display hHYD HYDFLJ11310 Hyperplastic discs protein homolog KIAA0896MGC57263 progestin induced protein Progestin-induced protein ubiquitin protein ligase E3 component n-recognin 5 ubiquitin-protein ligase; E3 identified by differential display; E3 ubiquitin-protein ligase UBR5; E3 ubiquitin-protein ligase, HECT domain-containing 1; HECT-type E3 ubiquitin transferase UBR5; hHYD; Hyperplastic discs protein homolog; Progestin-induced protein
Gene Aliases: DD5; EDD; EDD1; HYD; KIAA0896; UBR5
UniProt ID: (Human) O95071
Entrez Gene ID: (Human) 51366
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.