Invitrogen
Manufacturer Code:PA555715
Catalog # PIPA555715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UBR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase UBR2; E3 ubiquitin-protein ligase UBR2 N-recognin-2 Ubiquitin-protein ligase E3-alpha-2 Ubiquitin-protein ligase E3-alpha-II C6orf133 KIAA0349; N-recognin-2; RING-type E3 ubiquitin transferase UBR2; Ubiquitin-protein ligase E3-alpha-2; Ubiquitin-protein ligase E3-alpha-II
Gene Aliases: bA49A4.1; C6orf133; dJ242G1.1; dJ392M17.3; KIAA0349; UBR2
UniProt ID: (Human) Q8IWV8
Entrez Gene ID: (Human) 23304
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.