Proteintech
Manufacturer Code:188531AP150UL
Catalog # 50-173-2263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBR2 Fusion Protein Ag13446 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf133 KIAA0349 N recognin 2 RP3 392M17.3 UBR2; E3 ubiquitin-protein ligase UBR2; N-recognin-2; RING-type E3 ubiquitin transferase UBR2; ubiquitin ligase E3 alpha-II; Ubiquitin-protein ligase E3-alpha-2; Ubiquitin-protein ligase E3-alpha-II
Gene Aliases: 9930021A08Rik; AI462103; AW540746; bA49A4.1; C6orf133; dJ242G1.1; dJ392M17.3; E130209G04Rik; KIAA0349; mKIAA0349; UBR2
UniProt ID: (Human) Q8IWV8, (Mouse) Q8R130
Entrez Gene ID: (Human) 23304, (Mouse) 224826
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.