Abnova
Manufacturer Code:H00023304K
Catalog # 89-201-305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human UBR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human UBR2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: C6orf133 DKFZp686C08114 KIAA0349 MGC71112 RP3-392M17.3 bA49A4.1 dJ242G1.1 dJ392M17.3; E3 ubiquitin-protein ligase UBR2; N-recognin-2; RING-type E3 ubiquitin transferase UBR2; Ubiquitin-protein ligase E3-alpha-2; Ubiquitin-protein ligase E3-alpha-II
Gene Aliases: bA49A4.1; C6orf133; dJ242G1.1; dJ392M17.3; KIAA0349; UBR2
UniProt ID: (Human) Q8IWV8
Entrez Gene ID: (Human) 23304
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.