Invitrogen
Manufacturer Code:PA542370
Catalog # PIPA542370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human UBR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 alpha; E3 ubiquitin-protein ligase UBR1; E3a ligase; N-recognin-1; RING-type E3 ubiquitin transferase UBR1; ubiquitin ligase E3 alpha-I; Ubiquitin protein ligase E3 component n-recognin 1 JBS MGC142065 MGC142067; ubiquitin-protein ligase e3 componen n-recognin; ubiquitin-protein ligase E3-alpha; Ubiquitin-protein ligase E3-alpha-1; Ubiquitin-protein ligase E3-alpha-I
Gene Aliases: AI504731; JBS; RGD1562326; UBR1
UniProt ID: (Human) Q8IWV7, (Mouse) O70481
Entrez Gene ID: (Bovine) 522464, (Dog) 487521, (Rabbit) 100338337, (Human) 197131, (Horse) 100056497, (Mouse) 22222, (Rat) 499877, (Guinea pig) 100715624
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.