Invitrogen
Manufacturer Code:PA551974
Catalog # PIPA551974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UBL4A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: housekeeping protein DXS254E; ubiquitin-like 4; ubiquitin-like 4A; Ubiquitin-like 4A DX254E DXS254E G6PD GDX UBL4 GET5 MDY2 TMA24; Ubiquitin-like protein 4A; Ubiquitin-like protein GDX
Gene Aliases: DX254E; DXHXS254E; DXS254E; DXS254Eh; G6PD; GDX; GET5; MDY2; RGD1563983; TMA24; UBL4; UBL4A
UniProt ID: (Human) P11441, (Mouse) P21126, (Rat) B2GV38
Entrez Gene ID: (Human) 8266, (Mouse) 27643, (Rat) 293864
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.