Novus Biologicals
Manufacturer Code:NBP184956
Catalog # NBP184956
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VSTLKQLVSEKLNVPVRQQRLLFKGKALADGKRLSDYSIGPNSKLNLVVKPLEKVLLEEGEAQRLADSPPPQVWQLISKVLARHFSAADASRVLEQLQRDYERSLSRLTLDDIERLASRFLHPEVTETMEKG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DXS254EUbiquitin-like protein GDX G6PD GDXubiquitin-like 4 GET5 MDY2 TMA24 ubiquitin-like 4A ubiquitin-like protein 4A UBL4DX254E; ubiquitin-like 4; ubiquitin-like 4A; Ubiquitin-like protein 4A; Ubiquitin-like protein GDX
Gene Aliases: DX254E; DXS254E; G6PD; GDX; GET5; MDY2; TMA24; UBL4; UBL4A
UniProt ID: (Human) P11441
Entrez Gene ID: (Human) 8266
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.